Learn More
Jackson Immuno Research Labs 115-066-146
SDP
Supplier:  Jackson Immuno Research Labs 115066146
Encompass_Preferred
Biotin-SP-AffiniPure F(ab')2 Fragment Goat Anti-Mouse IgM, µ Chain Specific (min X Hu,Bov,Hrs Sr Prot)
Catalog No. NC0860150