Learn More
Sino Biological Recombinant Human Factor VII Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11506H08H20
Encompass_Preferred
A DNA sequence encoding the human F7 isoform B (P08709-2) (Met 1-Pro 444) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6916