Learn More
Sino Biological Recombinant Human NKp44/NCR2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11550H08H100
Encompass_Preferred
A DNA sequence encoding the human NCR2 (NP_004819.2) extracellular domain (Met 1-Pro 190) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6956