Learn More
Sino Biological Recombinant Human ESM1 Protein (His Tag)
Supplier:  Sino Biological 11578H08B100
Encompass_Preferred
A DNA sequence encoding the human ESM1 (NP_008967.1) (Met1-Arg184) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-7003