Learn More
Medchemexpress LLC 116-9e 100mg | 831217-43-7 | 100 MG

Supplier: Medchemexpress LLC HY116683100MG
116-9e (MAL2-11B) is a small-molecule inhibitor of the Hsp70 co-chaperone DNAJA1. It inhibits Simian Virus 40 (SV40) replication and DNA synthesis and suppresses tumor antigen (TAg) ATPase activity, making it suitable for research into chaperone-mediated viral replication and protein interactions.
- Molecular weight: 512.60 g/mol.
- Chemical formula: C31H32N2O5.
- CAS registry number: 831217-43-7.
- Purity: 98.28% (reported).
- Appearance: solid, white to yellow.
- Recommended form: 10 mM solution in DMSO (1 mL) or solid for reconstitution.
- Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (6 months) or -20°C (1 month).
- Pack size: 100 MG.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.