Learn More
Medchemexpress LLC 116-9e 10mg | 831217-43-7 | 512.60 Da | C31H32N2O5 | 10 MG

Supplier: Medchemexpress LLC HY11668310MG
116-9e (MAL2-11B) is a small-molecule inhibitor of the Hsp70 co-chaperone DNAJA1 that suppresses Simian Virus 40 (SV40) replication and DNA synthesis. It also inhibits tumor antigen (TAg) endogenous ATPase activity and TAg-mediated activation of Hsp70. Supplied as a white to yellow solid, the compound is intended for biochemical research and includes analytical documentation for quality verification.
- Inhibits DNAJA1 and reduces SV40 replication and DNA synthesis.
- Inhibits TAg endogenous ATPase activity.
- White to yellow solid suitable for biochemical assays.
- High purity (98.28%) as provided by supplier.
- Soluble in DMSO at 100 mg/mL for stock preparations.
- Powder storage: -20°C for long-term stability; in solution store at -80°C for extended storage.
- Analytical documentation available: COA, H NMR, LCMS, and SDS.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.