Learn More
Medchemexpress LLC 116-9e 25mg | 831217-43-7 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11668325MG
Encompass_Preferred
116-9e (MAL2-11B) is a small-molecule inhibitor of the Hsp70 co-chaperone DNAJA1 that inhibits Simian Virus 40 (SV40) replication and DNA synthesis. It is provided as a research-grade biochemical for in vitro and in vivo studies; recommended solvents and in vivo formulation protocols are available.
Catalog No. 50-002-26801