Learn More
Medchemexpress LLC 116-9e 50mg | 831217-43-7 | 512.60 | C31H32N2O5 | 50 MG

Supplier: Medchemexpress LLC HY11668350MG
116-9e (MAL2-11B) is a small-molecule inhibitor of the Hsp70 co-chaperone DNAJA1 and an inhibitor of Simian Virus 40 (SV40) replication and DNA synthesis. Supplied as a high-purity research reagent, it is intended for in vitro biochemical and cellular studies.
- High purity (98.28%) suitable for biochemical assays.
- Molecular formula C31H32N2O5 and molecular weight 512.60 g·mol-1.
- Soluble in DMSO; available as a 10 mM solution in DMSO for convenience.
- Offered in multiple solid pack sizes to fit different experimental scales.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.