Learn More
Sino Biological Recombinant Human USP7 Protein (His & GST Tag)
Supplier:  Sino Biological 11681H20B1100
Encompass_Preferred
A DNA sequence encoding full length of human USP7 (NP_003461.2) (Met 1-Asn 1102) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7155