Learn More
Sino Biological Influenza A H1N3 (A/duck/NZL/160/1976) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11685V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 529) of the influenza A Hemagglutinin (A/duck/NZL/160/1976 (H1N3)) (ABB20429.1) (HA1+HA2, uncleaved) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7191