Learn More
Sino Biological Influenza A H5N1 (A/Japanese white-eye/Hong Kong/1038/2006) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11694V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 530) of the influenza A hemagglutinin (A/Japanese white-eye/Hong Kong/1038 /2006 (H5N1)) (ABJ96775.1) (HA1+HA2, cleavage site mutated, RRRKR-TETR, uncleaved) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-7238