Learn More
Sino Biological Influenza A H5N3 (A/duck/Hokkaido/167/2007) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11696V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 342) of the influenza A Hemagglutinin H5N3 (A/duck/Hokkaido/167/2007) (BAG07130.2), termed as HA1, was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7248