Learn More
Sino Biological Recombinant Human CD42b/GP1BA Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11765H08H50
Encompass_Preferred
A DNA sequence encoding the human GP1BA (CAM97225.1) (Met1-Gly519) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7461