Learn More
eMolecules​ 1184-88-9 | Sodium pivalate | Ambeed | MFCD00064202 | 124.115 | C5H9NaO2 | 95.000 | [Na+].CC(C)(C)C([O-])=O | 100g | 591797585
Supplier:  eMolecules​ 591797585
Encompass_Preferred
Sodium pivalate | Ambeed | 1184-88-9 | MFCD00064202 | 124.115 | C5H9NaO2 | 95.000 | [Na+].CC(C)(C)C([O-])=O | 100g | 591797585
Catalog No. 50-004-89688