Learn More
Sino Biological Recombinant Human FSH Beta Protein (His Tag)
Supplier:  Sino Biological 11935H08H50
Encompass_Preferred
A DNA sequence encoding the human FSHB (NP_000501.1) (Met1-Glu129) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7627