Learn More
Sino Biological Recombinant Human Siglec-2/CD22 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 11958H02H50
Encompass_Preferred
A DNA sequence encoding the human CD22 (NP_001762.2) (Met1-Arg687) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-7644