Learn More
Sino Biological Recombinant Human GASP-1/WFIKKN2 Protein (His Tag)
Supplier:  Sino Biological 11998H08H50
Encompass_Preferred
A DNA sequence encoding the human WFIKKN2 (NP_783165.1) extracellular domain (Met 1-His 576) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7711