Learn More
Cayman Chemical 11dehydro Thromboxn B2 As 10mL
SDP
Supplier:  Cayman Chemical 41951710 ML
Encompass_Preferred
11-dehydro Thromboxane B2 Assay Buffer Concentrate (10X) has been formulated and tested to work with Cayman’s ELISA Kits. Detailed instructions for its use are contained in the respective Cayman ELISA Kit booklet
Catalog No. 50-294-3357