Learn More
Cayman Chemical 11nor9 ScarboxyHexahydro 1mg
SDP
Supplier:  Cayman Chemical 363561 MG
Encompass_Preferred
An analytical reference standard structurally similar to known phytocannabinoids; a metabolite of exo-THC and 9-hexahydrocannabinol; intended for research and forensic applications
Catalog No. 50-291-2106