Learn More
Cayman Chemical 11Z14Z17ZEIcosatrIen500ug
SDP
Supplier:  Cayman Chemical 17622500 UG
Encompass_Preferred
An esterified form of 11(Z),14(Z),17(Z)-eicosatrienoic acid; has been found in the lipid-producing microalgae Nannochloropsis; has been found in diesel-contaminated soils and residential wastewater
Catalog No. 50-296-5433