Learn More
MSC Thomas & Betts C10-10F #10 Stud, 12 to 10 AWG Compatible, Noninsulated, Crimp Connection, Standard Fork Terminal 0.38" Fork Width, Yellow, 0.82" Overall Length, 600 Volts, Copper Contacts
Supplier:  MSC 54064597
Encompass
Thomas & Betts C10-10F #10 Stud, 12 to 10 AWG Compatible, Noninsulated, Crimp Connection, Standard Fork Terminal 0.38" Fork Width, Yellow, 0.82" Overall Length, 600 Volts, Copper Contacts
Catalog No. 50-129-6034