Learn More
MSC Thomas & Betts 10RC-8F #8 Stud, 12 to 10 AWG Compatible, Partially Insulated, Crimp Connection, Standard Fork Terminal 0.38" Fork Width, Vinyl Insulation, Yellow, 1.09" Overall Length, 600 Volts, Copper Contacts
Supplier:  MSC 54064035
Encompass
Thomas & Betts 10RC-8F #8 Stud, 12 to 10 AWG Compatible, Partially Insulated, Crimp Connection, Standard Fork Terminal 0.38" Fork Width, Vinyl Insulation, Yellow, 1.09" Overall Length, 600 Volts, Copper Contacts
Catalog No. 50-129-6042