Learn More
Cayman Chemical 12 13DiHOME Fatty Acids and De
SDP
Supplier:  Cayman Chemical 1000983250 UG
Encompass_Preferred
A diol form of (±)12,13-EpOME; toxic to Sf21 cells expressing sEH and to lacZ-expressing control cells; rat spinal cord levels are increased following burn injury; BALF levels are elevated in humans following exposure to biodiesel exhaust and in exhaled breath condensate in patients with allergic asthma following allergen exposure; plasma levels are increased immediately following moderate-intensity exercise in mice and humans
Catalog No. 50-289-0371