Learn More
Cayman Chemical 12 13DiHOMEd4 GC MS Standards
SDP
Supplier:  Cayman Chemical 10009994250 UG
Encompass_Preferred
An internal standard for the quantification of 12(13)-DiHOME by GC- or LC-MS
Catalog No. 50-288-9186