Learn More
Novus Biologicals 12-230 kDa Separation Module for Peggy Sue or Sally Sue Systems, 480 Capillaries
SDP
Supplier:  Novus Biologicals SMS001
Encompass
Use with a Detection Module. Reagents for 480 data points includes two parts: Separation box with 5x boxes of Capillaries (96 capillaries per box), 5x Plates, Wash Buffer, Upper and Lower Running Buffers 3, Upper and Lower Running Buffer cups, Wash Buffer cup, 5x Manifold Cleaning Sponges, Separation Matrix 3, Stacking Matrix 2, 10X Sample Buffer, Antibody Diluent (Store at RT), plus 2x Standard Pack 3 clamshell each with 8xDTT and 8x FL master mix with 90 kDa System Control protein. (Store at 4 C).
Catalog No. NC1178176