Learn More
Novus Biologicals 12-230kDa Fluorescence Separation Module, 104 Capillaries
SDP
Supplier:  Novus Biologicals SMFL001
Encompass
Compatible with Jess™ systems. Includes separation box with Fluorescent Capillary Cartridges, 12-230kDa Pre-filled Plates (pre-filled with Separation Matrix 2, Stacking Matrix 2, Split Running Buffer 2 and Matrix Removal Buffer), Sample Buffer, Wash Buffer, plus Standard Pack 1 clamshell with DTT, FL Standard master mix with 29kDa System Control protein. Compatible with Detection module - sold separately.
Catalog No. NC2610323