Learn More
Medchemexpress LLC 12-Azido-dodecanoyl-OSu | 2489524-00-5 | C16H26N4O4 | 5 MG

Supplier: Medchemexpress LLC HY1518185MG
12-Azido-dodecanoyl-OSu is a click chemistry reagent featuring an azide group, serving as a hydrophobic bioconjugation linker.
- Contains an azide group
- Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
- Capable of ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN-containing molecules
- For research use only
- Appearance: Solid, white to off-white
- Purity: ≥97.0%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.