Learn More
Sklar Instruments Cannula Pipe Cleaners, Sklar 12 in., Cleaning Brush, Polypropylene, Disposable, PK50.
SDP
Supplier:  Sklar Instruments 101471
Encompass_Preferred
Cannula Pipe Cleaners, Sklar, 12 in., Cleaning Brush, Polypropylene, Disposable, PK50. Cannula Pipe Cleaners are typically used to clean secretions out of the inner cannula of a tracheostomy tube.
Catalog No. 50-109-4432