Learn More
Medchemexpress LLC 12-Deoxywithastramonolide | 60124-17-6 | 1 MG

Supplier: Medchemexpress LLC HYN71951MG
12-Deoxywithastramonolide is a principal bioactive compound from ashwagandha (Withania somnifera), known for its antioxidant and enzyme inhibitory properties. With a molecular weight of 470.60 and formula C28H38O6, it appears as a white to off-white solid, categorized under steroids from plant sources. For research use only, not human therapeutic use. Optimal storage: powder at -20°C (3 years) or 4°C (2 years); in solvent, -80°C (6 months) or -20°C (1 month). Shipping is stable at room temperature for less than two weeks.
- Principal bioactive compound from ashwagandha (W. somnifera)
- Possesses antioxidant and enzyme inhibitory effects
- Molecular weight of 470.60, formula C28H38O6
- White to off-white solid appearance
- Classified under steroids from plant origins
- For research use only
- Powder stable at -20°C (3 years) or 4°C (2 years)
- Solvent stable at -80°C (6 months) or -20°C (1 month)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.