Learn More
Medchemexpress LLC 12-Deoxywithastramonolide | 60124-17-6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN719510MG
Encompass_Preferred
12-Deoxywithastramonolide is a principal bioactive compound found in ashwagandha (W. somnifera). It possesses antioxidant and enzyme inhibitory effects, making it suitable for various biological research applications.
  • Naturally occurring compound found in ashwagandha (W. somnifera).
  • Exhibits antioxidant properties.
  • Possesses enzyme inhibitory capabilities.
Catalog No. 50-004-18837