Learn More
Sigma Aldrich Fine Chemicals Biosciences 12 Dichloroethane d4 99 at5G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 3965405G
Encompass
1 2-Dichloroethane-d4 is 1 2-dichloroethane with all the hydrogen atoms replaced by deuterium. Its synthesis and structure elucidation by neutron diffraction measurements has been reported.
Catalog No. 50-289-8761