Learn More
Medchemexpress LLC 12-Epinapelline | 110064-71-6 | 25 MG

Supplier: Medchemexpress LLC 25MGHYN2162
12-Epinapelline is a diterpene alkaloid isolated from Aconitum baikalense. This compound demonstrates anti-inflammatory activity and promotes the growth of colonies derived from fibroblast precursors. It is suitable for research use only.
- Exhibits anti-inflammatory activity
- Stimulates growth of colonies from fibroblast precursors
- Appears as a white to off-white solid
- Purity of 99.92%
- Molecular formula: C22H33NO3
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.