Learn More
Medchemexpress LLC 12-ethyl-4-methyl-11-(propan-2-yl)-1,8,10-triazatricyclo[7.4.0.0²,⁷]trideca-2(7),3,5,8,11-pentaen-13-one | 2641398-04-9 | MFCD35078192 | 99.9% | C16H19N3O | 10MG

Supplier: Medchemexpress LLC HY14519410MG
MrgprX2 antagonist-4 is a small-molecule antagonist of the human MRGPRX2 receptor intended for preclinical research into mast cell-driven inflammatory skin disorders and pseudo-allergic reactions. It is supplied for in vitro and in vivo studies and is not for human therapeutic use.
- High purity suitable for research (99.9% reported).
- Molecular formula C16H19N3O; molecular weight 269.34 g·mol⁻¹.
- Soluble in DMSO; in vitro solubility ~5 mg/mL with warming and sonication.
- In vivo formulation: 10% DMSO in corn oil (clear solution ≥0.5 mg/mL).
- Intended for laboratory research use only, not for clinical use.
- Useful tool compound for studies of mast cell degranulation and skin inflammation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.