Learn More
Medchemexpress LLC 12-Hydroxystearic ac 250mg
SDP
Supplier:  Medchemexpress LLC 250MGHYW127329
Encompass_Preferred
12-Hydroxystearic acid is a structurally simple and cost-effective low molecular weight organogelator and its metal salts and derivatives find roles in many important applications[1][2]
Catalog No. 50-002-05224