Learn More
DANCO PACKAGING PRODUCTS LLC 12 INCH IMPULSE HEAT SEALER
Supplier:  DANCO PACKAGING PRODUCTS LLC 15MP12
Encompass_Preferred
Impulse Heat Sealer, Length: 12 in., Variable Seal Time: 0.2 to 1.3 sec., Uses 380W and plugs into any wall outlet, it creates 1/16 in.. seal, it will seal polyethylene or polypropylene materials up to 5mil. in thickness
Catalog No. NC9159355