Learn More
Medchemexpress LLC 12-ketochenodeoxycholic acid | 2458-08-4 | MFCD00134876 | 99.8% | 406.56 g/mol | C24H38O5 | 1 ML

Supplier: Medchemexpress LLC HYW40393310MM/1ML
12-ketochenodeoxycholic acid is a bile acid derivative and metabolic precursor of cholodeoxycholic acid supplied for research use. It is available as a white to off-white solid or as a ready-to-use 10 mM solution in DMSO, and is commonly used in studies of bile acid metabolism and gut microbiota metabolites.
- Purity 99.8%.
- CAS number 2458-08-4.
- Chemical formula C24H38O5; molecular weight 406.56 g/mol.
- Available as solid and as 10 mM solution in DMSO (1 mL).
- Appearance white to off-white solid.
- Storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months) or -20 °C (1 month).
- Solubility DMSO 100 mg/mL; ultrasonic assistance may be required.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.