Learn More
Cayman Chemical 12-oxo PhytodIenoIc AcId 100UG
SDP
Supplier:  Cayman Chemical 88520100 UG
Encompass_Preferred
A biologically active, immediate precursor of 7-epi jasmonic acid; plays a role independent of jasmonic acid activity in mediating resistance to pathogens and pests
Catalog No. 50-289-4442