Learn More
Enzo Life Sciences 12(R)-Hydroxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (HETE) (50µg). CAS: 82337-46-0
SDP
Supplier:  Enzo Life Sciences BMLH1120050
Encompass_Preferred
Biosynthesis in psoriatic skin and corneal epithelial cells via cytochrome P-450. More potent than 12(S)-HETE in inducing human leukocyte chemokinesis and aggregation. Proinflammatory effects in human skin. Inhibits renal, cardiac and corneal Na+,K+-ATPase. Reduces intraocular pressure. Alternative name: 12(R)-HETE. Formulation: Liquid. Solution in ethanol. Purity: ≥99%. Long Term Storage: -80°C.
Catalog No. 50-201-1021