Learn More
Cayman Chemical 12 Shydroxy16HeptadecynoI 10mg
SDP
Supplier:  Cayman Chemical 3157010 MG
Encompass_Preferred
12(S)-hydroxy-16-Heptadecynoic acid is a mechanism-based inhibitor of CYP450 ω-hydroxylase.{1051} It inhibits prostaglandin ω-hydroxylase with a Ki value of 1.8 µM.{1051}
Catalog No. 50-299-0892