Learn More
WATSON BIO LAB INC 1200 uL Extra Long (130 mm) Hyper-Filter Tip, Graduated, Sterilized, 960 tips(96 tips * 10 racks), PC system rack
Supplier:  WATSON BIO LAB INC 1251200S
Encompass_Preferred
Extra-long pipette tips designed for volumes up to 1,200 µL
Ideal for accessing deep vessels such as centrifuge tubes
Hydrophobic porous filters help reduce the risk of liquid entering the pipette
Graduated markings allow quick visual confirmation of aspirated volume
Supplied sterile in a convenient 96-tip rack
Catalog No. NC2283843