Learn More
Sino Biological Recombinant Human SMOC-1 Protein (His Tag)
Supplier:  Sino Biological 12026H08H100
Encompass_Preferred
A DNA sequence encoding the human SMOC1 isoform 1 (NP_001030024.1) (Met 1-Val 435) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7726