Learn More
Sino Biological Recombinant Human Pepsinogen C/PGC Protein (His Tag)
Supplier:  Sino Biological 12072H07E50
Encompass_Preferred
A DNA sequence encoding the mature form of human PGC (P20142-1) (Ile153-Ile239) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-7776