Learn More
eMolecules​ 1211589-41-1 | 5-Bromo-2-trifluoromethyl-isonicotinic acid methyl ester | Apollo Scientific | MFCD16657895 | 284.032 | C8H5BrF3NO2 | 95.000 | COC(=O)c1cc(ncc1Br)C(F)(F)F | 5g | 633426544
Supplier:  eMolecules​ 633426544
Encompass_Preferred
5-Bromo-2-trifluoromethyl-isonicotinic acid methyl ester | Apollo Scientific | 1211589-41-1 | MFCD16657895 | 284.032 | C8H5BrF3NO2 | 95.000 | COC(=O)c1cc(ncc1Br)C(F)(F)F | 5g | 633426544
Catalog No. 50-001-61153