Learn More
Sino Biological Recombinant Human Siglec-3/CD33 Protein (His Tag), Amine-Labeled Biotinylated, HPLC-verified
Supplier:  Sino Biological 12238H08HB20
Encompass_Preferred
A DNA sequence encoding the human CD33 (AAH28152.1) (Met1-His259) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-161-7908