Learn More
Cayman Chemical 1234 6 7 8 9OctaCLOrodIb 10mg
SDP
Supplier:  Cayman Chemical 3859810 MG
Encompass_Preferred
A polychlorinated dibenzodioxin; increases EROD activity in isolated rat liver microsomes (ED50 = 750 UG/kg); induces hepatic fatty vacuolization in rats at 50 UG/kg; has been found in animal feed, freshwater fish, ash/soil from open burn sites, and in the air near municipal waste incinerators
Catalog No. 50-291-8973