Learn More
Medchemexpress LLC 123B9 | 2225868-19-7 | 99.7% | 1321.79 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP105795MG
Encompass_Preferred
123B9 is a tumor-homing agent that acts as a potent and selective EphA2 agonist with a Kd value of 4.0 μM. It selectively targets the EphA2 tyrosine kinase receptor ligand-binding domain and does not significantly inhibit the ligand binding domains of the closely related EphA3 and EphA4 receptors.
  • Potent and selective EphA2 agonist
  • Tumor-homing agent
  • Selectively targets the EphA2 tyrosine kinase receptor ligand-binding domain
Catalog No. 50-004-23688