Learn More
Refrigiwear Inc. 123CR BLACK SIZE 12 PLATINUM LEATHER BOOT
SDP
Supplier:  Refrigiwear Inc. 123CRBLK120
Encompass_Preferred
Don't let foot fatigue, discomfort or injury keep you from getting the job done with these full grain leather boots. The non-slip outsole and safety toe keep you safe, while the exclusive VitaComfort® system keeps your feet comfortable and dry. Comfort Rating: -35�F to 15�F 1000g Insulation 8" Buffalo leather upper Vibram® Montana with IceTrek® outsole ASTM/CSA composite safety toe Welted construction CSA puncture resistant plate Rubber toe guard Waterproof VitaComfort® anti-fatigue and gravity-fed moisture management system ESR/Electrical hazard compliant    
Catalog No. NC3867065