Learn More
Sino Biological Recombinant Human Rack1 Protein (His & MBP Tag), HPLC-verified
Supplier:  Sino Biological 12498H10E100
Encompass_Preferred
A DNA sequence encoding the human GNB2L1 (P63244) (Met 1-Arg 317) was fused with an N-terminal polyhistidine-tagged MBP tag at the N-terminus.
Catalog No. 50-161-8041