Learn More
Alkali Scientific 125 mL Erlenmeyer Flask Bi-directional Transfer Cap with Silicon Tube (50cm 1/8" ID 1/4" OD), Male CPC Connector with Female Sealing Cap, Vent Filter (0.22 μm Φ42mm), without Flask, Sterile, 1/pk, 4/cs
SDP
Supplier:  Alkali Scientific EFC400
Encompass_Preferred
Compatible with the Erlenmeyer Flask and involved in the suspended culture and medium preparation or storage in cell biology and microbiology. For aseptic transfer of culture medium and cell culture solution. One end of the pipeline is connected to the MPC series connector, which increases the convenience and safety in use. The male connector and the sealing cap are tightly connected to prevent accidental disconnection, and the connector can be rotated to reduce pipeline distortion. (50cm 1/8" ID 1/4" OD), Male CPC Connector with Female Sealing Cap, Vent Filter (0.22 μm Φ42mm)
Catalog No. 50-302-8862