Learn More
Alkali Scientific Bulk Pipet Tips, 1250ul, Bagged, 1000/pk
SDP
Supplier:  Alkali Scientific BT1250
Encompass
Bulk Pipet Tips by ASI are ultra-clear and universal fit to most pipettes. Made from 100% polypropylene, low-retention plastic, these tips ensure accurate sample results during liquid handling. The Pipet Tip line is packaged Rnase/Dnase, pyrogen free and available in both sterile and non-sterile bags of 1000 tips.
Catalog No. 50-238-9288